Merck
  • High Color-Rendering Index and Stable White Light-Emitting Diodes by Assembling Two Broadband Emissive Self-Trapped Excitons.
High Color-Rendering Index and Stable White Light-Emitting Diodes by Assembling Two Broadband Emissive Self-Trapped Excitons.
Advanced materials (Deerfield Beach, Fla.) (2020-11-24)
Zhuangzhuang Ma, Zhifeng Shi, Dongwen Yang, Yawen Li, Fei Zhang, Lintao Wang, Xu Chen, Di Wu, Yongtao Tian, Yu Zhang, Lijun Zhang, Xinjian Li, Chongxin Shan
ABSTRACT
White light-emitting diodes (WLEDs) are promising next-generation solid-state light sources. However, the commercialization route for WLED production suffers from challenges in terms of insufficient color-rendering index (CRI), color instability, and incorporation of rare-earth elements. Herein, a new two-component strategy is developed by assembling two broadband emissive materials with self-trapped excitons (STEs) for high CRI and stable WLEDs. The strategy addresses effectively the challenging issues facing current WLEDs. Based on first-principles thermodynamic calculations, copper-based ternary halides composites, CsCu2 I3 @Cs3 Cu2 I5 , are synthesized by a facile one-step solution approach. The composites exhibit an ideal white-light emission with a cold/warm white-light tuning and a robust stability against heat, ultraviolet light, and environmental oxygen/moisture. A series of cold/warm tunable WLEDs is demonstrated with a maximum luminance of 145 cd m-2 and an external quantum efficiency of 0.15%, and a record high CRI of 91.6 is achieved, which is the highest value for lead-free WLEDs. Importantly, the fabricated device demonstrates an excellent operation stability in a continuous current mode, exhibiting a long half-lifetime of 238.5 min. The results promise the use of the hybrids of STEs-derived broadband emissive materials for high-performance WLEDs.
Sigma-Aldrich
Copper(I) iodide, 99.999% trace metals basis