Merck
  • Nonenzymatic Glucose Sensors Based on Copper Sulfides: Effect of Binder-Particles Interactions in Drop-Casted Suspensions on Electrodes Electrochemical Performance.
Nonenzymatic Glucose Sensors Based on Copper Sulfides: Effect of Binder-Particles Interactions in Drop-Casted Suspensions on Electrodes Electrochemical Performance.
Sensors (Basel, Switzerland) (2021-02-04)
Julia Mazurków, Anna Kusior, Marta Radecka
ABSTRACT
The constant progress in novel nanomaterials synthesis has contributed to the rapid development of nonenzymatic glucose sensors. For working electrodes preparation, drop casting proved to be the most convenient and thus most widely applied method. However, appropriate interpretation of obtained electrochemical signal requires in-depth knowledge of limitations related to this technique. In this study, we prepared solutions based on commonly reported polymers for nanostructures immobilization and investigated their influence on copper sulfides distribution on the electrode. Characterization of suspensions properties and behavior of particles during droplet drying revealed that nonionic polyvinylpyrrolidone (PVP) was favorable for electrodes modification with copper sulfides in comparison with Nafion and chitosan. It ensured homogeneity of the suspension as well as the uniform coverage of the electrode surface with particles, what resulted in increased active surface area and, therefore, higher signal from glucose addition. On the other hand, when cationic chitosan was used as a binder, suspensions were agglomerated and, within dry deposits, a coffee-ring effect was observed. Appropriate adjustment of material and polymer interactions led to enhanced electrode electrochemical performance.
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis