Merck
  • Aging-Associated miR-217 Aggravates Atherosclerosis and Promotes Cardiovascular Dysfunction.
Aging-Associated miR-217 Aggravates Atherosclerosis and Promotes Cardiovascular Dysfunction.
Arteriosclerosis, thrombosis, and vascular biology (2020-08-28)
Virginia G de Yébenes, Ana M Briones, Inmaculada Martos-Folgado, Sonia M Mur, Jorge Oller, Faiz Bilal, María González-Amor, Nerea Méndez-Barbero, Juan Carlos Silla-Castro, Felipe Were, Luis J Jiménez-Borreguero, Fátima Sánchez-Cabo, Héctor Bueno, Mercedes Salaices, Juan Miguel Redondo, Almudena R Ramiro
ABSTRACT
microRNAs are master regulators of gene expression with essential roles in virtually all biological processes. miR-217 has been associated with aging and cellular senescence, but its role in vascular disease is not understood. Approach and Results: We have used an inducible endothelium-specific knock-in mouse model to address the role of miR-217 in vascular function and atherosclerosis. miR-217 reduced NO production and promoted endothelial dysfunction, increased blood pressure, and exacerbated atherosclerosis in proatherogenic apoE-/- mice. Moreover, increased endothelial miR-217 expression led to the development of coronary artery disease and altered left ventricular heart function, inducing diastolic and systolic dysfunction. Conversely, inhibition of endogenous vascular miR-217 in apoE-/- mice improved vascular contractility and diminished atherosclerosis. Transcriptome analysis revealed that miR-217 regulates an endothelial signaling hub and downregulates a network of eNOS (endothelial NO synthase) activators, including VEGF (vascular endothelial growth factor) and apelin receptor pathways, resulting in diminished eNOS expression. Further analysis revealed that human plasma miR-217 is a biomarker of vascular aging and cardiovascular risk. Our results highlight the therapeutic potential of miR-217 inhibitors in aging-related cardiovascular disease.
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder