- Identification of transcripts regulated by CUG-BP, Elav-like family member 1 (CELF1) in primary embryonic cardiomyocytes by RNA-seq.
Identification of transcripts regulated by CUG-BP, Elav-like family member 1 (CELF1) in primary embryonic cardiomyocytes by RNA-seq.
CUG-BP, Elav-like family member 1 (CELF1) is a multi-functional RNA binding protein that regulates pre-mRNA alternative splicing in the nucleus, as well as polyadenylation status, mRNA stability, and translation in the cytoplasm [1]. Dysregulation of CELF1 has been implicated in cardiomyopathies in myotonic dystrophy type 1 and diabetes [2-5], but the targets of CELF1 regulation in the heart have not been systematically investigated. We previously demonstrated that in the developing heart CELF1 expression is restricted to the myocardium and peaks during embryogenesis [6-8]. To identify transcripts regulated by CELF1 in the embryonic myocardium, RNA-seq was used to compare the transcriptome of primary embryonic cardiomyocytes following siRNA-mediated knockdown of CELF1 to that of controls. Raw data files of the RNA-seq reads have been deposited in NCBI's Gene Expression Omnibus [9] under the GEO Series accession number GSE67360. These data can be used to identify transcripts whose levels or alternative processing (i.e., alternative splicing or polyadenylation site usage) are regulated by CELF1, and should provide insight into the pathways and processes modulated by this important RNA binding protein during normal heart development and during cardiac pathogenesis.