Merck
  • A neuron-specific enhancer targets expression of the gonadotropin-releasing hormone gene to hypothalamic neurosecretory neurons.
A neuron-specific enhancer targets expression of the gonadotropin-releasing hormone gene to hypothalamic neurosecretory neurons.
Molecular endocrinology (Baltimore, Md.) (1995-04-01)
D B Whyte, M A Lawson, D D Belsham, S A Eraly, C T Bond, J P Adelman, P L Mellon
ABSTRACT
The molecular mechanisms specifying gene expression in individual neurons of the mammalian central nervous system have been difficult to study due to the cellular complexity of the brain and the absence of cultured model systems representing differentiated central nervous system neurons. We have developed clonal, differentiated, neuronal tumor cell lines of the hypothalamic GnRH-producing neurons by targeting tumorigenesis in transgenic mice. These cells (GT1 cells) provide a model system for molecular studies of GnRH gene regulation. Here we present the identification and characterization of a neuron-specific enhancer responsible for directing expression of the rat GnRH gene in GT1 hypothalamic neurons. This approximately 300 base pair (bp) upstream region (-1571 to -1863) confers enhancer activity to a short -173-bp GnRH promoter or to a heterologous promoter only in GT1 cells. The enhancer is bound by multiple GT1 nuclear proteins over its entire length. Deletion of more than 30 bp from either end dramatically reduces activity, and even large internal fragments carrying seven of the eight DNAse I-protected elements show decreased activity. Scanning replacement mutations demonstrate that several of the internal elements are required for activity of the enhancer. Thus, the GnRH gene is targeted to hypothalamic neurons by a complex multicomponent enhancer that relies on the interaction of multiple nuclear-protein binding enhancer elements.
Sigma-Aldrich
GT1-7 Mouse Hypothalamic GnRH Neuronal Cell Line, GT1-7 cells can be used as an in vitro model of GnRH-secreting neurons of the hypothalamus.