Merck
  • Controlled release formulations of metribuzin: release kinetics in water and soil.
Controlled release formulations of metribuzin: release kinetics in water and soil.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2010-04-20)
Jitendra Kumar, Keyath Nisar, N A Shakil, Suresh Walia, Rajender Parsad
ABSTRACT
Controlled release (CR) formulations of metribuzin in Polyvinyl chloride [(PVC) (emulsion)], carboxy methyl cellulose (CMC), and carboxy methyl cellulose-kaolinite composite (CMC-KAO), are reported. Kinetics of its release in water and soil was studied in comparison with the commercial formulation (75 DF). Metribuzin from the commercial formulation became non-detectable after 35 days whereas it attained maxima between 35-49 days and became non-detectable after 63 days in the developed products. Amongst the CR formulations, the release in both water and soil was the fastest in CMC and slowest in PVC. The CMC-KAO composite reduced the rate of release as compared to CMC alone. The diffusion exponent (n value) of metribuzin in water and soil ranged from 0.515 to 0.745 and 0.662 to 1.296, respectively in the various formulations. The release was diffusion controlled with half release time (t(1/2)) from different controlled release matrices of 12.98 to 47.63 days in water and 16.90 to 51.79 days in soil. It was 3.25 and 4.66 days, respectively in the commercial formulation. The period of optimum availability of metribuzin in water and soil from controlled released formulations ranged from 15.09 to 31.68 and 17.99 to 34.72 days as against 5.03 and 8.80 days in the commercial formulation.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Pricing and availability is not currently available.
Supelco
Metribuzin, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Pricing and availability is not currently available.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Pricing and availability is not currently available.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Pricing and availability is not currently available.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Pricing and availability is not currently available.