Merck
  • Ghrelin O-acyltransferase (GOAT) has a preference for n-hexanoyl-CoA over n-octanoyl-CoA as an acyl donor.
Ghrelin O-acyltransferase (GOAT) has a preference for n-hexanoyl-CoA over n-octanoyl-CoA as an acyl donor.
Biochemical and biophysical research communications (2009-06-09)
Hideko Ohgusu, Kaori Shirouzu, Yuki Nakamura, Yoshiki Nakashima, Takanori Ida, Takahiro Sato, Masayasu Kojima
ABSTRACT
Ghrelin is a peptide hormone in which serine 3 is modified by n-octanoic acid through GOAT (ghrelin O-acyltransferase). However, the enzymological properties of GOAT remain to be elucidated. We analyzed the in vitro activity of GOAT using the recombinant enzyme. Unexpectedly, although the main active form of ghrelin is modified by n-octanoic acid, GOAT had a strong preference for n-hexanoyl-CoA over n-octanoyl-CoA as an acyl donor. Moreover, a four-amino acid peptide derived from the N-terminal sequence of ghrelin can be modified by GOAT, indicating that these four amino acids constitute the core motif for substrate recognition by the enzyme.
Product Description
Avanti
06:0 Coenzyme A, Avanti Research - A Croda Brand 870706P, powder
Sigma-Aldrich
Octanoyl coenzyme A lithium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Hexanoyl coenzyme A trilithium salt hydrate, ≥85%