Merck
  • Regioselective Arylboration of Isoprene and Its Derivatives by Pd/Cu Cooperative Catalysis.
Regioselective Arylboration of Isoprene and Its Derivatives by Pd/Cu Cooperative Catalysis.
Journal of the American Chemical Society (2017-06-06)
Kevin B Smith, M Kevin Brown
ABSTRACT
A method for the regioselective arylboration of isoprene and its derivatives is presented. These reactions allow for the synthesis of useful building blocks from simple components. Through these studies, an unusual additive effect with DMAP has been uncovered that allows for altered reactivity and the formation of quaternary carbon centers. The utility of this method is demonstrated toward the formal synthesis of mesembrine.
Sigma-Aldrich
QPhos Pd G3
Sigma-Aldrich
QPhos Pd G3 ChemBeads