
trans-β-Farnesene

Synonym(s):
trans-7,11-Dimethyl-3-methylene-1,6,10-dodecatriene
Empirical Formula (Hill Notation):
C15H24
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing