
Rhodamine 110 chloride

Empirical Formula (Hill Notation):
C20H14N2O3 · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing