![Propamocarb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/402/250/a3369d77-4a18-4552-9124-e4629a3455fb/640/a3369d77-4a18-4552-9124-e4629a3455fb.png)
Propamocarb
![Propamocarb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/402/250/a3369d77-4a18-4552-9124-e4629a3455fb/640/a3369d77-4a18-4552-9124-e4629a3455fb.png)
Empirical Formula (Hill Notation):
C9H20N2O2
CAS Number:
Molecular Weight:
188.27
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2