Skip to Content
Merck
PIPES sodium salt ≥99% (titration)

PIPES sodium salt

PIPES sodium salt ≥99% (titration)
Synonym(s):
1,4-Piperazinediethanesulfonic acid sodium salt, Piperazine-N,N′-bis(2-ethanesulfonic acid) sodium salt
Linear Formula:
C8H17N2O6S2Na
CAS Number:
Molecular Weight:
324.35
EC Number:

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
pK Value
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing