
Pimobendan for system suitability

Synonym(s):
Pimobendan, 4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-3(2H)-pyridazinone
Empirical Formula (Hill Notation):
C19H18N4O2
CAS Number:
Molecular Weight:
334.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing