
PCB No 52

Synonym(s):
2,2′,5,5′-Tetrachlorobiphenyl
Empirical Formula (Hill Notation):
C12H6Cl4
CAS Number:
Molecular Weight:
291.99
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing