
Octanoic acid-1-13C

Synonym(s):
Caprylic acid-1-13C
Linear Formula:
CH3(CH2)613CO2H
CAS Number:
Molecular Weight:
145.20
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing