
N-Demethylazithromycin

Synonym(s):
3′-N-Desmethylazithromycin
Empirical Formula (Hill Notation):
C37H70N2O12
CAS Number:
Molecular Weight:
734.96
Products
Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2