Skip to Content
Merck
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt ≥97% (HPLC), powder

N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt

N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt ≥97% (HPLC), powder
Synonym(s):
Bucladesine sodium salt, Dibutyryl cAMP sodium salt, Dibutyryl cyclic-AMP sodium salt
Linear Formula:
C18H23N5O8PNa
CAS Number:
Molecular Weight:
491.37
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Showing 1-2 of 2