
Methyl all-cis-5,8,11,14,17-eicosapentaenoate

Synonym(s):
cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester
Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2CH3
CAS Number:
Molecular Weight:
316.48
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing