Skip to Content
Merck
Imipenem Pharmaceutical Secondary Standard; Certified Reference Material

Imipenem

Imipenem Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Imipenem monohydrate, (5R,6S)-6-[(1R)-1-Hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid monohydrate, Primaxin monohydrate, Tienam monohydrate
Empirical Formula (Hill Notation):
C12H17N3O4S · H2O
CAS Number:
Molecular Weight:
317.36
EC Number:

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing