
IgG1−FITC Isotype Control from murine myeloma

Synonym(s):
IgG1 Isotype Control from murine myeloma
Products
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing