
Hypaconitine

Synonym(s):
Hypaconitine, 16,16-trimethoxy-4-(methoxymethyl)-20-methyl-,8-acetate benzoate,(1-α,6-α,14-α,15-α,16-β)-aconitane-8,13,14,15-tetraol
Empirical Formula (Hill Notation):
C33H45NO10
CAS Number:
Molecular Weight:
615.71
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing