Glyceryl tri(oleate-1-13C)
Synonym(s):
Triolein-carboxyls-13C3
Linear Formula:
[CH3(CH2)7CH=CH(CH2)713CO2CH2]2CH[O213C(CH2)7CH=CH(CH2)7CH3]
CAS Number:
Molecular Weight:
888.41
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing