
(−)-Gallocatechin

Synonym(s):
(2S,3R)-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol
Empirical Formula (Hill Notation):
C15H14O7
CAS Number:
Molecular Weight:
306.27
Products
Product Category
Brand
Formula Weight
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing