
Fmoc-D-beta-homoproline

Synonym(s):
Fmoc-′D-β-homoproline′
Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
Products
Product Category
Brand
Formula Weight
Physical Form
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing