
Diisobutyl fumarate

Linear Formula:
(CH3)2CHCH2O2CCH=CHCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
228.28
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing