Skip to Content
Merck
Dibromomethane-d2 ≥99 atom % D, ≥99% (CP), contains copper as stabilizer

Dibromomethane-d2

Dibromomethane-d2 ≥99 atom % D, ≥99% (CP), contains copper as stabilizer
Synonym(s):
Methylene-d2 bromide
Linear Formula:
CD2Br2
CAS Number:
Molecular Weight:
175.85

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing