Skip to Content
Merck
Cidofovir Related Compound B certified reference material, pharmaceutical secondary standard

Cidofovir Related Compound B

Cidofovir Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
1-[(S)-3-Hydroxy-2-(O,O-diethylphosphonomethoxy)propyl]cytosine hydrochloride
Empirical Formula (Hill Notation):
C12H22N3O6P · HCl
CAS Number:
Molecular Weight:
371.75

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade