
Amifostine

Synonym(s):
Amifostine trihydrate, 2-(3-Aminopropyl)aminoethyl phosphorothioate, WR2721
Empirical Formula (Hill Notation):
C5H15N2O3PS · 3 H2O
CAS Number:
Molecular Weight:
268.27
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing