
5′ -Cytidylic Acid

Synonym(s):
Cytidine 5′-monophosphate, 5′-CMP, 5′-Cytidylic acid, C-5′-P
Empirical Formula (Hill Notation):
C9H14N3O8P
CAS Number:
Molecular Weight:
323.20
Products
Feature
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2