
5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt

Synonym(s):
Magenta-GlcA, Magenta™ glucuronide
Empirical Formula (Hill Notation):
C14H13BrClNO7 · C6H13N
CAS Number:
Molecular Weight:
521.79
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing