2-Chloro-3-nitrobenzoic acid
Linear Formula:
ClC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
201.56
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing