Skip to Content
Merck
(2-Bromovinyl)trimethylsilane approx. 90% trans, 98%

(2-Bromovinyl)trimethylsilane

(2-Bromovinyl)trimethylsilane approx. 90% trans, 98%
Synonym(s):
β-(Trimethylsilyl)vinyl bromide, 1-Bromo-2-(trimethylsilyl)ethylene
Linear Formula:
BrCH=CHSi(CH3)3
CAS Number:
Molecular Weight:
179.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing