
(1-Methyl-5-nitroimidazol-2-yl)methanol

Synonym(s):
2-Hydroxymethyl-1-methyl-5-nitro-1H-imidazole
Empirical Formula (Hill Notation):
C5H7N3O3
CAS Number:
Molecular Weight:
157.13
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing