Skip to Content
Merck
All Photos(17)

Key Documents

ZRB1415

Sigma-Aldrich

Anti-Nucleolin Antibody, clone 5D13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein C23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 76.61 kDa
observed mol wt ~110 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NCL(4691)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 5D13 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Nucleolin. It targets an epitope within 16 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Nucleolin.

Application

Anti-Nucleolin, clone 5D13 ZooMAb, Cat. No. ZRB1415, is a recombinant Rabbit monoclonal antibody that targets Nucleolin and is tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Nucleolin in A431 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Nucleolin in HeLa, HUVEC, as A431 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Nucleolin in human small intestine tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Nucleolin (UniProt: P19338; also known as Protein C23) is encoded by the NCL gene (Gene ID: 4691) in human. Nucleolin is a multifunctional phosphoprotein that is ubiquitously distributed in the nucleolus, nucleus and cytoplasm of the cell. It is one of the most abundant non-ribosomal proteins that accounts for about 10% of the protein content within the nucleolus. It is found associated with intra-nucleolar chromatin and pre-ribosomal particles. It plays important roles in various steps of ribosomal synthesis, including the transcription of rDNA repeats, the modification and processing of pre-rRNA, the assembly of pre-ribosomal particles and nuclear-cytoplasmic transport of ribosomal proteins and subunits. Nucleolin contains three structural and multifunctional domains: the N-terminal domain contains several acidic stretches; two to four RNA-binding domains (RNA recognition motifs or RRM) in the central region; and a glycine/arginine-rich domain (GAR) domain at the C-terminus. Nucleolin is shown to be highly phosphorylated. It undergoes extensive phosphorylation by casein kinase 2 at interphase and by CDC2 during mitosis. It has been reported that Nucleolin phosphorylation at interphase is correlated with active rRNA transcription. Although the mammalian Nucleolin has a predicted molecular weight of about 77 kDa, its apparent molecular weight lies between 100 and 110 kDa due to highly phosphorylated amino acids in the N-terminal domain. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Tajrishi, MM et al., (2011). Commun. Integr. Biol. 4(3); 267-275).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.03 mg/mL after reconstitution at 25μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service