Skip to Content
Merck
All Photos(1)

Key Documents

SLW1200/12D

Pyrex® Kjeldahl flask

plain neck

Synonym(s):

Pyrex, SciLabware 1200/12D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

Pyrex flask
round bottom
single-neck

feature

plain neck

packaging

pack of 10 ea

manufacturer/tradename

1200/12D
SciLabware

H

325 mm

bulb diam.

101 mm

capacity

500 mL
500 mL

neck I.D. × O.D.

30 mm × 34 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Suitable for the determination of nitrogen etc.
  • Manufactured from Pyrex borosilicate glass
  • Withstands high temperatures
  • Resistant to chemical corrosion

Features and Benefits

• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Used for the determination of nitrogen by the Kjeldahl method•

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service