Merck
HPLCによるアミノ酸(極性、中性、酸性)の分析、Ascentis® Si
HPLCによるアミノ酸(極性、中性、酸性)の分析、Ascentis® Si application for HPLC
材料
分析カラム
製品番号
詳細
価格
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Si, 10 cm x 2.1 mm I.D., 5 μm particles (581508-U)
mobile phase
[A] 100 mM ammonium formate, pH 3.0, with formic acid; [B] water; [C] acetonitrile
gradient
5% A constant; 10% B, 85% C to 16% B, 79% C in 20 min
flow rate
0.3 mL/min
column temp.
35 °C
detector
ESI (+), full scan
injection
2 μL
sample
as indicated, in 10:90, (50 mM ammonium formate/formic acid, (pH 3.0):acetonitrile