コンテンツへスキップ
Merck
  • Proteomic Signature Reveals Modulation of Human Macrophage Polarization and Functions Under Differing Environmental Oxygen Conditions.
Proteomic Signature Reveals Modulation of Human Macrophage Polarization and Functions Under Differing Environmental Oxygen Conditions.
Molecular & cellular proteomics : MCP (2017-09-10)
Magali Court, Graciane Petre, Michèle El Atifi, Arnaud Millet
要旨
Macrophages are innate immune cells which can react to a large number of environmental stimuli thanks to a high degree of plasticity. These cells are involved in a variety of tissue functions in homeostasis, and they play essential roles in pathological contexts. Macrophages' activation state, which determines their functional orientation, is strongly influenced by the cellular environment. A large body of macrophage literature is devoted to better defining polarizations from a molecular viewpoint. It is now accepted that a multidimensional model of polarization is needed to grasp the broad phenotype repertoire controlled by environmental signals. The study presented here aimed, among other goals, to provide a molecular signature of various polarizations in human macrophages at the protein level to better define the different macrophage activation states. To study the proteome in human monocyte-derived macrophages as a function of their polarization state, we used a label-free quantification approach on in-gel fractionated and LysC/Trypsin digested proteins. In total, 5102 proteins were identified and quantified for all polarization states. New polarization-specific markers were identified and validated. Because oxygen tension is an important environmental parameter in tissues, we explored how environmental oxygen tension, at either atmospheric composition (18.6% O
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
5-カルボキシテトラメチルローダミン