コンテンツへスキップ
Merck
  • microRNA-21a-5p/PDCD4 axis regulates mesenchymal stem cell-induced neuroprotection in acute glaucoma.
microRNA-21a-5p/PDCD4 axis regulates mesenchymal stem cell-induced neuroprotection in acute glaucoma.
Journal of molecular cell biology (2017-06-29)
Wenru Su, Zuohong Li, Y Jia, Yingting Zhu, Wenjia Cai, Peixing Wan, Yingying Zhang, Song Guo Zheng, Yehong Zhuo
要旨
Mesenchymal stem cells (MSCs) have been demonstrated to have promising therapeutic benefits for a variety of neurological diseases; however, the underlying mechanisms are poorly understood. Here, we showed that intravitreal infusion of MSCs promoted retinal ganglion cell (RGC) survival in a mouse model of acute glaucoma, with significant inhibition of microglial activation, production of TNF-α, IL-1β, and reactive oxygen species, as well as caspase-8 and caspase-3 activation. In vitro, MSCs inhibited both caspase-8-mediated RGC apoptosis and microglial activation, partly via the action of stanniocalcin 1 (STC1). Furthermore, we found that microRNA-21a-5p (miR-21) and its target, PDCD4, were essential for STC1 production and the neuroprotective property of MSCs in vitro and in vivo. Importantly, miR-21 overexpression or PDCD4 knockdown augmented MSC-mediated neuroprotective effects on acute glaucoma. These data highlight a previously unrecognized neuroprotective mechanism by which the miR-21/PDCD4 axis induces MSCs to secrete STC1 and other factors that exert neuroprotective effects. Therefore, modulating the miR-21/PDCD4 axis might be a promising strategy for clinical treatment of acute glaucoma and other neurological diseases.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
MISSION® esiRNA, targeting human PDCD4
Sigma-Aldrich
MISSION® esiRNA, targeting human STC1