コンテンツへスキップ
Merck
  • TGF-β participates choroid neovascularization through Smad2/3-VEGF/TNF-α signaling in mice with Laser-induced wet age-related macular degeneration.
TGF-β participates choroid neovascularization through Smad2/3-VEGF/TNF-α signaling in mice with Laser-induced wet age-related macular degeneration.
Scientific reports (2017-08-31)
Xiaolei Wang, Wei Ma, Song Han, Zhaoyang Meng, Lu Zhao, Yi Yin, Yanling Wang, Junfa Li
要旨
Choroidal neovascularization(CNV) is the most severe complication in Age-related macular degeneration(AMD) and the most common cause of irreversible blindness in the elderly in developed world. The aim of this study was to identify the effect of transforming growth factor-β(TGF-β) and Smad2/3-VEGF/TNF-α signaling on CNV angiopoiesis, and to explore TGF-β inhibitors on the development of CNV in a CNV mouse model. Fundus fluorescein angiography(FFA) was used to evaluate the laser-induced CNV formation. The histology of CNV lesions stained with hematoxylin-eosin(HE) was obtained. The immunofluorescent staining was performed to determine TGF-β protein expression. The expressions of TGF-β, phosphorylated Smad2/3, VEGF and TNF-α were determined by using Western blot analysis. The CNV areas were analyzed by using fluorescein stain on RPE/choroid-sclera flat mounts. We found the levels of TGF-β protein expression increasingly reached the peak till 3rd week during the CNV development. The protein levels of VEGF and TNF-α also increased significantly in CNV mice, which were inhibited by a synthetic TGF-β inhibitor LY2157299 or a natural TGF-β inhibitor Decorin. The phosphorylated Smad2/3 levels increased significantly in CNV mice, but this response was profoundly suppressed by the TGF-β inhibitors. Here we have demonstrated that TGF-β/Smad signaling plays an important role in Laser-induced CNV formation through down-regulation of VEGF and TNF-α expressions, suggesting TGF-β inhibitors may provide an alternative to traditional methods in wet AMD treatment.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
デコリン ウシ関節軟骨由来, salt-free, lyophilized powder