コンテンツへスキップ
Merck
  • [Expression of CysLTR-1 and CysLTR-2 in adenoid tissues from children with adenoid hypertrophy].
[Expression of CysLTR-1 and CysLTR-2 in adenoid tissues from children with adenoid hypertrophy].
Zhongguo dang dai er ke za zhi = Chinese journal of contemporary pediatrics (2015-03-12)
Mei-Hua Zhu, Min Liang, Zhi-Jian Wang, Yan Shen, Hong-Yan Wen, Jin-Ming Zhai, Xiao-Bin Yang
要旨
To examine the expression of cysteinyl leukotriene receptor-1 (CysLTR-1) and cysteinyl leukotriene receptor-2 (CysLTR-2) in the adenoid tissues from children with adenoid hypertrophy (AH) and to explore the role of leukotrienes in the pathogenesis of AH. Sixty children with AH who were treated by adenoidectomy and/or tonsillectomy were classified into two groups: simple AH and AH plus allergic rhinitis (n=30 each). Twenty children who underwent tonsillectomy due to recurrent purulent tonsillitis were selected as the control group. The expression of CysLTR-1 and CysLTR-2 in the excised tonsil and/or adenoid tissues was determined by immunofluorescence histochemical labeling and integrated optical density measurement. The expression of CysLTR-1 and CysLTR-2 in the adenoid and tonsil tissues increased significantly in both the simple AH group and AH plus allergic rhinitis group compared with the control group (P<0.01). The expression of CysLTR-1 and CysLTR-2 in the AH plus allergic rhinitis group increased more significantly compared with the simple AH group (P<0.01). CysLTR-1 and CysLTR-2 are highly expressed in the adenoid tissues from children with AH, suggesting that leukotrienes are involved in the pathogenesis of AH.