コンテンツへスキップ
Merck
  • The use of (5Z)-4-bromo-5-(bromomethylene)-2(5H)-furanone for controlling acid mine drainage through the inhibition of Acidithiobacillus ferrooxidans biofilm formation.
The use of (5Z)-4-bromo-5-(bromomethylene)-2(5H)-furanone for controlling acid mine drainage through the inhibition of Acidithiobacillus ferrooxidans biofilm formation.
Bioresource technology (2015-03-25)
Yang Zhao, Peng Chen, Wenbin Nan, Dejuan Zhi, Ronghui Liu, Hongyu Li
要旨
The aim of this study was to determine whether acid mine drainage (AMD) production can be decreased by (5Z)-4-bromo-5-(bromomethylene)-2(5H)-furanone (furanone C-30) in the presence of Acidithiobacillus ferrooxidans (A. ferrooxidans). The effects of furanone C-30 on A. ferrooxidans biofilm production were determined by crystal violet staining and confocal laser scanning microscopy (CLSM). Biofilm-related gene expression was investigated using real-time RT-PCR. Finally, the effects of furanone C-30 on AMD production were evaluated. The results show that furanone C-30 inhibits the production of extracellular polymeric substances (EPS) and biofilm formation and significantly down-regulates the expression of biofilm-related genes. The decreased EPS production led to reduced pentlandite attachment and biofilm formation on pentlandite. Furthermore, the dissolution of both nickel and copper were inhibited by furanone C-30 without new acid formation. This study provides a promising biochemical method to control AMD.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
(Z-)-4-ブロモ-5-(ブロモメチレン)-2(5H)-フラノン, ≥97.0% (GC)