コンテンツへスキップ
Merck
  • Rapid renal alpha-1 antitrypsin gene induction in experimental and clinical acute kidney injury.
Rapid renal alpha-1 antitrypsin gene induction in experimental and clinical acute kidney injury.
PloS one (2014-05-23)
Richard A Zager, Ali C M Johnson, Kirsten B Frostad
要旨
Alpha-1-antitrypsin (AAT) is a hepatic stress protein with protease inhibitor activity. Recent evidence indicates that ischemic or toxic injury can evoke selective changes within kidney that resemble a hepatic phenotype. Hence, we tested the following: i) Does acute kidney injury (AKI) up-regulate the normally renal silent AAT gene? ii) Does rapid urinary AAT excretion result? And iii) Can AAT's anti-protease/anti-neutrophil elastase (NE) activity protect injured proximal tubule cells? CD-1 mice were subjected to ischemic or nephrotoxic (glycerol, maleate, cisplatin) AKI. Renal functional and biochemical assessments were made 4-72 hrs later. Rapidly following injury, 5-10 fold renal cortical and isolated proximal tubule AAT mRNA and protein increases occurred. These were paralleled by rapid (>100 fold) increases in urinary AAT excretion. AKI also induced marked increases in renal cortical/isolated proximal tubule NE mRNA. However, sharp NE protein levels declines resulted, which strikingly correlated (r, -0.94) with rising AAT protein levels (reflecting NE complexing by AAT/destruction). NE addition to HK-2 cells evoked ∼95% cell death. AAT completely blocked this NE toxicity, as well as Fe induced oxidant HK-2 cell attack. Translational relevance of experimental AAT gene induction was indicated by ∼100-1000 fold urinary AAT increases in 22 AKI patients (matching urine NGAL increases). We conclude: i) AKI rapidly up-regulates the renal cortical/proximal tubule AAT gene; ii) NE gene induction also results; iii) AAT can confer cytoprotection, potentially by blocking/reducing cytotoxic NE accumulation; and iv) marked increases in urinary AAT excretion in AKI patients implies clinical relevance of the AKI- AAT induction pathway.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ヒトリポカリン-2 / NGAL ELISAキット, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse Lcn2 / Neutrophil Gelatinase-associated Lipocalin ELISA Kit
Sigma-Aldrich
Rat Lcn2 / Neutrophil Gelatinase-associated Lipocalin ELISA Kit