コンテンツへスキップ
Merck
  • Circulating levels of the vasoactive peptide urotensin II in patients with acute coronary syndrome and stable coronary artery disease.
Circulating levels of the vasoactive peptide urotensin II in patients with acute coronary syndrome and stable coronary artery disease.
Peptides (2014-03-20)
Hamood Al Kindi, Anouar Hafiane, Zhipeng You, Isabella Albanese, Louise Pilote, Jacques Genest, Adel Schwertani
要旨
Urotensin II (UII) is a vasoactive peptide with various roles in cardiovascular physiology and pathophysiology. There is an accumulating evidence implicating UII in atherosclerosis and coronary artery disease, making it an important target in acute coronary syndrome (ACS). In this study, we sought to determine the plasma levels of UII in ACS patients within 48 h of clinical presentation and after a 12-week recovery period. We compared them to patients with stable coronary artery disease (CAD) and a control group of normolipidemic subjects without known CAD. Using a highly sensitive ELISA technique, we measured plasma UII in 27 ACS patients, 26 stable CAD patients and 22 age-matched controls. ACS patients had significantly elevated plasma UII during the first 48 h of clinical presentation compared to stable CAD patients and controls. We also found significant positive correlations between UII and CRP and with triglycerides and a significant negative correlation between UII and EF. There was no correlation with LDL-C. In conclusion, plasma UII levels were elevated in patients with acute coronary syndrome, particularly immediately after clinical presentation. This suggests an upregulation of UII expression in ACS.