コンテンツへスキップ
Merck
  • B-cell maturation antigen is a promising target for adoptive T-cell therapy of multiple myeloma.
B-cell maturation antigen is a promising target for adoptive T-cell therapy of multiple myeloma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2013-01-25)
Robert O Carpenter, Moses O Evbuomwan, Stefania Pittaluga, Jeremy J Rose, Mark Raffeld, Shicheng Yang, Ronald E Gress, Frances T Hakim, James N Kochenderfer
要旨
Multiple myeloma is a usually incurable malignancy of plasma cells. New therapies are urgently needed for multiple myeloma. Adoptive transfer of chimeric antigen receptor (CAR)-expressing T cells is a promising new therapy for hematologic malignancies, but an ideal target antigen for CAR-expressing T-cell therapies for multiple myeloma has not been identified. B-cell maturation antigen (BCMA) is a protein that has been reported to be selectively expressed by B-lineage cells including multiple myeloma cells. Our goal was to determine if BCMA is a suitable target for CAR-expressing T cells. We conducted an assessment of BCMA expression in normal human tissues and multiple myeloma cells by flow cytometry, quantitative PCR, and immunohistochemistry. We designed and tested novel anti-BCMA CARs. BCMA had a restricted RNA expression pattern. Except for expression in plasma cells, BCMA protein was not detected in normal human tissues. BCMA was not detected on primary human CD34(+) hematopoietic cells. We detected uniform BCMA cell-surface expression on primary multiple myeloma cells from five of five patients. We designed the first anti-BCMA CARs to be reported and we transduced T cells with lentiviral vectors encoding these CARs. The CARs gave T cells the ability to specifically recognize BCMA. The anti-BCMA-CAR-transduced T cells exhibited BCMA-specific functions including cytokine production, proliferation, cytotoxicity, and in vivo tumor eradication. Importantly, anti-BCMA-CAR-transduced T cells recognized and killed primary multiple myeloma cells. BCMA is a suitable target for CAR-expressing T cells, and adoptive transfer of anti-BCMA-CAR-expressing T cells is a promising new strategy for treating multiple myeloma.