コンテンツへスキップ
Merck
  • Protein expression of programmed death 1 ligand 1 and ligand 2 independently predict poor prognosis in surgically resected lung adenocarcinoma.
Protein expression of programmed death 1 ligand 1 and ligand 2 independently predict poor prognosis in surgically resected lung adenocarcinoma.
OncoTargets and therapy (2014-04-22)
Yang Zhang, Lei Wang, Yuan Li, Yunjian Pan, Rui Wang, Haichuan Hu, Hang Li, Xiaoyang Luo, Ting Ye, Yihua Sun, Haiquan Chen
要旨
The clinicopathologic characteristics of tumors expressing programmed death (PD-1) ligands (PD-Ls) PD-L1 or PD-L2 and their associations with common driver mutations in lung adenocarcinoma are not clearly defined, despite the progression of anti-PD-1/PD-L1 immunotherapy. PD-L1 and PD-L2 expression was measured by immunohistochemistry in 143 surgically resected lung adenocarcinomas and was correlated with clinical variables, histologic subtypes, and the mutational status of EGFR, KRAS, HER2, and ALK. Positive PD-L1 expression was significantly associated with more advanced T status, N status, and pathologic stage. Histologically, lung adenocarcinomas with positive PD-L1 staining were less likely to be adenocarcinoma in situ or minimally invasive adenocarcinoma and more likely to have solid predominant subtype. Both PD-L1 expression (odds ratio =1.984, 95% confidence interval =1.010-3.894; P=0.047) and PD-L2 expression (odds ratio =2.328, 95% confidence interval =1.201-4.512; P=0.012) were independent predictors of poor overall survival. When the combined PD-L expression and pathologic stage were used together to predict overall survival, the concordance index increased to 0.763, and the Akaike information criteria value decreased to 356.08. We defined the clinicopathologic features of lung adenocarcinomas with high expression of PD-L1 and PD-L2. We further demonstrated the role of PD-L expression as a useful prognostic marker for lung adenocarcinoma.