コンテンツへスキップ
Merck
  • Quantitative RT-PCR assay for MUC3 and VEGF mRNA in renal clear cell carcinoma: relationship with nuclear grade and prognosis.
Quantitative RT-PCR assay for MUC3 and VEGF mRNA in renal clear cell carcinoma: relationship with nuclear grade and prognosis.
Urology (2003-10-11)
Xavier Leroy, Valerie Gouyer, Charles Ballereau, Farid Zerimech, Guillemette Huet, Marie-Christine Copin, Jean-Pierre Aubert, Nicole Porchet
要旨
To investigate, by real-time quantitative reverse transcriptase-polymerase chain reaction, the expression of MUC3 and vascular endothelial growth factor (VEGF) to correlate them with histologic parameters and with prognosis. Human mucins are large O-glycoproteins expressed in epithelial tissues. Deregulation of mucin genes has been demonstrated in several epithelial neoplasms. In the kidney, MUC3 is expressed in normal convoluted tubules and in renal clear cell carcinoma. Twenty-six renal clear cell carcinoma specimens were studied. For all tumors, samples of normal and tumor kidney were frozen. After RNA extraction, using ultracentrifugation through a cesium chloride cushion, VEGF and MUC3 mRNA were analyzed by real-time quantitative reverse transcriptase-polymerase chain reaction. The pathologic parameters included Fuhrman nuclear grade and TNM stage. All follow-up data were available. The median level of MUC3 and VEGF expression was greater in tumor areas compared with normal areas (P < 0.002 and P < 0.001, respectively). The MUC3 tumor/normal tissue expression ratio was greater in nuclear grade 3 tumor than in low grades (grade 1-2; P < 0.005). No statistically significant relationship was found with the prognosis for MUC3 and VEGF in our study. The results of this study demonstrate that MUC3 and VEGF are overexpressed in renal clear cell carcinoma, and the MUC3 expression ratio is greater in nuclear grade 3 than in grades 1 and 2 (low grades) tumor. These findings suggest the implication of MUC3 in renal carcinogenesis.