コンテンツへスキップ
Merck
  • Efficient synthesis and structure-activity relationship of honokiol, a neurotrophic biphenyl-type neolignan.
Efficient synthesis and structure-activity relationship of honokiol, a neurotrophic biphenyl-type neolignan.
Bioorganic & medicinal chemistry letters (2004-04-28)
Tomoyuki Esumi, Gouki Makado, Haifeng Zhai, Yasuhiro Shimizu, Yasuhide Mitsumoto, Yoshiyasu Fukuyama
要旨
Honokiol, a biphenyl-type neolignan, which shows the remarkable neurotrophic effect in primary cultured rat cortical neurons, has been effectively synthesized in 21% yield over 14 steps starting from 5-bromosalicylic acid and p-hydroxybenzoic acid by utilizing Pd-catalyzed Suzuki-Miyaura coupling reaction as a key step. Additionally, the structure-activity relationship between neurite outgrowth-promoting activity and its O-methylated and/or its hydrogenated analogues was examined in the primary cultures of fetal rat cortical neurons, suggesting that 5-allyl and 4'-hydroxyl groups are essential for affecting the neurotrophic activity of honokiol.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
5-ブロモサリチル酸, technical grade, 90%