コンテンツへスキップ
Merck
  • Assessment of the genotoxic and mutagenic properties of Himatanthus articulatus bark extracts used as phytotherapeutic drug in the Amazon.
Assessment of the genotoxic and mutagenic properties of Himatanthus articulatus bark extracts used as phytotherapeutic drug in the Amazon.
Journal of ethnopharmacology (2013-03-27)
Sebastião de O Rebouças, Juliana da Silva, Ricardo S Bertoni, Natalia Decker, Marcela Silva Dos Santos, Raíssa R Rossatto, Dione S Corrêa, Alexandre B F Ferraz
要旨
Himatanthus articulatus (Apocynaceae) is a plant native to the Amazon, popularly used to treat external ulcers, tumors, inflammations, cancer, syphilis and malaria. To investigate the in vivo genotoxic and mutagenic potential of this plant, using the comet assay and the micronucleus test. Female and male adult mice were treated with 500 mg/kg, 1000 mg/kg or 2000 mg/kg of Himatanthus articulatus aqueous or ethanolic bark extracts by gavage for two consecutive days. In addition, blood slides were exposed to hydrogen peroxide (ex vivo) to evaluate the anticlastogenic effect using the comet assay. The HPLC analyses indicated plumieride as the main constituent of both extracts from Himatanthus articulatus barks. No differences between genders were observed. Micronuclei were observed only in the group treated with the highest dose of both extracts. Conversely, lower doses of these extracts showed protective effects to DNA against damage induced by hydrogen peroxide, indicating an important antigenotoxic effect. The toxicological evaluation indicated that the extracts are non-genotoxic and reduce the clastogenic damage induced by hydrogen peroxide. In part, this result can be atributted to the phytochemical profile of Himatanthus articulatus, which presents iridoids and phenolic compounds.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Plumieride, ≥95% (LC/MS-ELSD)