コンテンツへスキップ
Merck
  • Pharmacokinetics of single and multiple oral doses of arbidol in healthy Chinese volunteers.
Pharmacokinetics of single and multiple oral doses of arbidol in healthy Chinese volunteers.
International journal of clinical pharmacology and therapeutics (2013-02-09)
Yaxin Sun, Xiaojing He, Feng Qiu, Xu Zhu, Mingming Zhao, Jesse Li-Ling, Xianying Su, Limei Zhao
要旨
Arbidol is licensed in Russia and China for prophylaxis and treatment of influenza A and B. This study was to assess the pharmacokinetics of single and multiple doses of arbidol in healthy Chinese volunteers. This was a single-center, open-label, two-phase study conducted in 12 subjects. In single-dose phase, subjects were randomized to receive single doses of 0.2, 0.4 and 0.8 g of arbidol in a crossover design with a 7-day washout period between administration. In the multiple-dose phase, subjects received 0.2 g 3 times a day for 7 days. Serial blood samples were collected at predefined time points. Plasma concentrations were determined with a validated HPLC method. Safety assessments were conducted throughout the study. After administration of single doses of 0.2, 0.4 and 0.8 g, geometric mean estimates for arbidol Cmax were 0.70, 1.24, and 2.16 mg/l and the mean of AUClast were 3.27, 5.81 and 12.72 mg×h/l, respectively. The AUClast and Cmax showed dose proportionality. After administration of multiple doses, the mean of Cmax,ss of arbidol was 0.41 mg/l and the mean accumulation ratio is ~ 1.12. Compared with single-dose phase, arbidol exhibited lower Cmax and prolonged plasma concentration profiles. In healthy Chinese subjects, single dosing of arbidol resulted in linear plasma pharmacokinetics. Arbidol exhibited little accumulation with repeated administration. Compared with single doses, multiple oral doses showed somewhat different pharmacokinetics and tissue distribution patterns. Sex did not appear to affect the pharmacokinetic properties of arbidol.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Arbidol hydrochloride, ≥98% (HPLC)