コンテンツへスキップ
Merck
  • Early clinical observations in prospectively followed patients with fungal meningitis related to contaminated epidural steroid injections.
Early clinical observations in prospectively followed patients with fungal meningitis related to contaminated epidural steroid injections.
Annals of internal medicine (2012-11-28)
Thomas M Kerkering, Marissa L Grifasi, Anthony W Baffoe-Bonnie, Ekta Bansal, Dorothy C Garner, Jean A Smith, Deborah D Demicco, Charles J Schleupner, Rabia A Aldoghaither, Vipul A Savaliya
要旨
Chinese translation Administration of epidural steroid injections (ESIs) with contaminated methylprednisolone resulted in an outbreak of fungal meningitis in many locations in the United States. To characterize early clinical findings and initial response to treatment. Case series with standardized observation studied from 4 October to 31 October 2012. An 800-bed hospital in Virginia. 172 patients who presented to the hospital with exposure to contaminated ESI. Standardized approach to screening, case definition, treatment, and data collection. Clinical findings, cerebrospinal fluid (CSF) values, magnetic resonance imaging (MRI), serum and CSF voriconazole concentrations, and clinician assessment of response to therapy. Of 172 patients presenting to the hospital who had had ESI, 131 had lumbar puncture because of symptoms or signs consistent with central nervous system disease. Twenty-five (19%) had neutrophilic meningitis. All were started on voriconazole therapy alone. Three patients developed stroke during treatment. Ten patients had arachnoiditis, another had an epidural abscess, and 9 had urine retention. Fifteen continued to receive voriconazole, and 10 were switched to amphotericin B. Cerebrospinal fluid leukocyte counts began to decrease by day 13 of treatment. Findings on MRI included ventriculitis, leptomeningeal enhancement, infarction, hemorrhage, and arachnoiditis. Serum voriconazole levels varied, and CSF concentrations of voriconazole were approximately 50% those of serum. Exserohilum rostratum and Cladosporium species have been cultured. This is an observational study of an evolving outbreak. Not all exposed patients presented for evaluation. Follow-up is too short to determine final outcomes. Meningitis after receipt of contaminated ESI has been diagnosed in many exposed patients presenting to 1 hospital. Most patients have improved on receipt of empirical voriconazole therapy. The full natural history and long-term sequelae of this infection are currently unknown. None.
材料
製品番号
ブランド
製品内容
Methylprednisolone acetate, European Pharmacopoeia (EP) Reference Standard